Learn More
Skytec™ TRC715 Safety Gloves
Product Code. p-200110066
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
10 pairs
Product Code. 30274509
Brand: Skytec™ SKG00016ED
Only null left
Add to basket
Add to basket
Outstanding cut resistance allowing protection in high risk applications. Breathable, long lasting and flexible glove providing excellent grip in oily conditions.
  • Recycled Polyester
  • HPPE on the inside provides comfort and protection
  • Fully encapsulated steel with recycled polyester on the outside strengthens the protection against burrs and sharp edges
  • Micro foam nitrile coating gives exceptional grip while allowing warm air and moisture from inside to escape
Specifications
Certifications/Compliance (Europe) CE II, UKCA, ANSI/ISEA 105-2016: Cut A5, EN 388:2016 4X43E, EN 388:2016 + A1:2018 4X43E
Coating Material Foam Nitrile
Color Green, Black
Cut Resistance Rating Level E
Hand Ambidextrous
Latex Free Yes
Material Steel Fiber
Product Type Cut-resistant Gloves
Size Small
EN388 Standard 4X43E
Liner Material HPPE, Recycled Polyester
Length (Metric) 230 mm