missing translation for 'onlineSavingsMsg'
Learn More

Trehalulose, TRC

Product Code. 30180248
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180248 25 mg 25mg
30179852 100 mg 100mg
30179807 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180248 Supplier Toronto Research Chemicals Supplier No. TRCT20748025MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51411-23-5
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.3
Synonym 1-O-alpha-D-Glucopyranosyl-D-fructose, 1-O-alpha-D-Glucosylfructose, Mildear;Mildear 75, Trehalulose, Vitalose
IUPAC Name (3S,4R,5R)-3,4,5,6-tetrahydroxy-1-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexan-2-one
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H22O11/c13-1-4(15)7(17)8(18)5(16)3-22-12-11(21)10(20)9(19)6(2-14)23-12/h4,6-15,17-21H,1-3H2/t4-,6-,7-,8-,9-,10+,11-,12+/m1/s1
Formula Weight 342.1162
Chemical Name or Material Trehalulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.