Learn More
uvex Trend BOA™ Black/Gray/Red PU/PU 69082 Low Shoe - Width 11
Product Code. 17203301 Shop All Uvex Products
Change view
Click to view available options
Size:
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Unit Size:
1 pair
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17203301 38 1 pair
17213301 39 1 pair
17223301 40 1 pair
17233301 41 1 pair
17243301 42 1 pair
17253301 43 1 pair
17263301 44 1 pair
17273301 45 1 pair
17283301 46 1 pair
17293301 47 1 pair
17203311 48 1 pair
17213311 49 1 pair
17223311 50 1 pair
17233311 51 1 pair
17243311 52 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 17203301 Supplier uvex Supplier No. 6908238
  • ONLINE EXCLUSIVE PRICE
    117.00 €
    107.18 € / 1 pairs
    Save 9.82 €
    8% Off
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
Uvex 2 Trend Perforated Shoe with BOA™ Fit System 6908.2 S1 P SRC
  • Sporty, S1 P perforated safety shoe with wide fit
  • Breathable perforated microvelour
  • Made from synthetic materials, suitable for people allergic to chrome
  • All sole materials free of silicones, plasticizers and other paint wetting impairment substances
  • BOA™ Fit System Dial for custom comfort that's consistent all day
  • Meets ESD specification with volume resistance under 35 megohms
  • Reliable protection for wearers
  • Steel toe cap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Reflective details for enhanced visibility
  • Uvex Climazone technology lining, upper and removable comfortable climatic insole with a moisture transport system
    • Helps maintain foot temperature for optimum wearer comfort
  • Soft padding on collar and tongue
  • Removable anti-static comfortable insole with moisture transport system and additional shock absorption around heel and forefoot
  • Article No. 6908.2
  • Standard EN ISO 20345:2011 S1 P SRC
  • Sizes: 38 to 52
TRUSTED_SUSTAINABILITY
Specifications
Color Black, Gray, Red
Material Microvelour
Size 38
Sole Material Dual-density polyurethane (PU/PU)
Toe Material Steel
Disposable Reusable
Certifications/Compliance (Europe) EN ISO 20345:2011
Gender Unisex
Fit Standard Width
Model Uvex 2
Latex Free Yes
Waterproof No
EN ISO 20345 S1
Slip Resistance Rating SRC
Product Type ESD Safety Shoe