missing translation for 'onlineSavingsMsg'
Learn More

Treosulfan, TRC

Product Code. 30003905
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003905 5 mg 5mg
30010472 10 mg 10mg
30012314 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003905 Supplier Toronto Research Chemicals Supplier No. TRCT2209055MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 299-75-2
Molecular Formula C6H14O8S2
Molecular Weight (g/mol) 278.3
Synonym (2S,3S)-1,2,3,4-Butanetetrol 1,4-Dimethanesulfonate, [S-(R*,R*)]-1,2,3,4-Butanetetrol 1,4-Dimethanesulfonate, (2S,3S)-Threitol 1,4-Dimethanesulfonate, L-Threitol 1,4-Bis(methanesulfonate), NSC 39069, Ovastat, Threosulphan, Trecondi, Treosulphan, Tresulfan, (2S,3S)-2,3-Dihydroxybutane-1,4-diyl Dimethanesulfonate
IUPAC Name [(2S,3S)-2,3-dihydroxy-4-methylsulfonyloxybutyl] methanesulfonate
SMILES CS(=O)(=O)OC[C@H](O)[C@@H](O)COS(=O)(=O)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C6H14O8S2/c1-15(9,10)13-3-5(7)6(8)4-14-16(2,11)12/h5-8H,3-4H2,1-2H3/t5-,6-/m0/s1
Formula Weight 278.013
Chemical Name or Material Treosulfan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.