Learn More
Beschreibung
- Laboratory Worktops and Weighing Surfaces: Erlab worktops are designed to provide a durable, chemically resistant and easy-to-clean working surface for ductless enclosures, filtering storage cabinets and laboratory benches. Each surface is dimensioned to fit the corresponding Erlab equipment exactly.
- Materials Selected for Laboratory Use
- High-resistance materials suitable for daily contact with laboratory chemicals
- Smooth, non-porous surface that is quick and easy to decontaminate
- Resistant to scratching, impact and routine cleaning agents
- Stainless Steel versions available for applications requiring maximum chemical and biological resistance
- Segmented and weighing versions available for specific handling requirements
- Designed for Safe Handling: The worktop geometry contributes to the containment performance of the enclosure by maintaining correct airflow at the working surface. Segmented versions allow easy removal for cleaning or for passing equipment through, while weighing surfaces provide a stable, vibration-damped supPort for precision balances.
- Key Benefits:
- Chemically resistant, easy-to-clean working surface.
- Exact fit with the corresponding Erlab equipment.
- Long service life under intensive laboratory use.
- Options available for weighing and segmented configurations.
- Contributes to correct airflow and containment performance.
Spezifikation
Spezifikation
| For Use With (Equipment) | Captair Ductless Filtering Fume Hoods |
| Product Type | Work Surface |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?