Learn More
Emma Safety Footwear Treviso Safety Shoes
Product Code. 15908151
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15908151
Brand: Emma Safety Footwear MM10509239
Only null left
Add to basket
Add to basket
The foot-hugging fit provides the wearer with a naturally light and airy on-foot feel. Water resistant uppers and a woven insole make the shoes virtually penetration proof. The 100% nitrile rubber outsole repels liquids, harmful or otherwise, and provides extra grip on almost all surfaces.
  • O3-Work shoe without steel toecap with sole plate
  • Full grain leather construction
  • Nitrile Rubber Heat Resistant Outsole
  • Metal-free
  • Chrome-free lining
  • ESD certified
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20347:2012
Closure Type Laces
Color Black
Design Treviso
Liner Material Hydro-Tec™ Sanitized Silver & Wellness leather
Material Leather
Size 39