Learn More
Emma Safety Footwear Treviso Safety Shoes
The foot-hugging fit provides the wearer with a naturally light and airy on-foot feel. Water resistant uppers and a woven insole make the shoes virtually penetration proof. The 100% nitrile rubber outsole repels liquids, harmful or otherwise, and provides extra grip on almost all surfaces.
Brand:  Emma Safety Footwear MM10509241
Additional Details : Weight : 1.43500kg
 View more versions of this product
Product Code. 15928151
  • 163.24 € / 1 pair
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
Description
Description
  • O3-Work shoe without steel toecap with sole plate
  • Full grain leather construction
  • Nitrile Rubber Heat Resistant Outsole
  • Metal-free
  • Chrome-free lining
  • ESD certified
  • Sanitized
Specifications
Specifications
EN ISO 20347:2012
Black
Hydro-Tec™ Sanitized Silver & Wellness leather
41
Laces
Treviso
Leather