missing translation for 'onlineSavingsMsg'
Learn More

Tri-Ammonium Citrate, Certified AR for analysis, Fisher Chemical™

Product Code. 10528800 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10528800 250 g 250g
10538800 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10528800 Supplier Fisher Chemical Supplier No. A/4082/50
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Tri-Ammonium Citrate, Certified AR for analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3458-72-8
Molecular Formula C6H17N3O7
Molecular Weight (g/mol) 243.22
MDL Number 36406
InChI Key YWYZEGXAUVWDED-UHFFFAOYSA-N
IUPAC Name triammonium 2-hydroxypropane-1,2,3-tricarboxylate
SMILES [NH4+].[NH4+].[NH4+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Melting Point 185°C
Color White
pH 7 to 8
Physical Form Solid
Quantity 250 g
Formula Weight 243.22g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Citric acid, triammonium salt
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.