missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyl(3-methyl-2-butenyl)tin, 95%

Product Code. 11309066
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11309066 1 g 1g
11319066 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309066 Supplier Thermo Scientific Alfa Aesar Supplier No. L20214.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 53911-92-5 | C17H36Sn | 359.185 g/mol

Tri-n-butyl(3-methyl-2-butenyl)tin is used in prenylation of ethyl propiolate. It is also involved in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butyl(3-methyl-2-butenyl)tin is used in prenylation of ethyl propiolate. It is also involved in chemical research.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53911-92-5
Molecular Formula C17H36Sn
Molecular Weight (g/mol) 359.185
MDL Number MFCD01863649
InChI Key XEFRYQPTIWSVGI-UHFFFAOYSA-N
Synonym tributyl 3-methylbut-2-en-1-yl stannane, tributyl 3-methyl-2-butenyl tin, tributylprenylstannane, acmc-1am9p, tri-n-butyl 3-methyl-2-butenyl tin, tributyl 3-methylbut-2-enyl stannane, stannane, tributyl 3-methyl-2-butenyl, tributyl 3-methylbut-2-en-1-yl ;stannane
PubChem CID 2734722
IUPAC Name tributyl(3-methylbut-2-enyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)CC=C(C)C

Specifications

Density 1.042
Boiling Point 283°C
Flash Point >110°C (230°F)
Refractive Index 1.488
Quantity 1 g
UN Number UN2788
Beilstein 3588742
Solubility Information Immiscible with water.
Formula Weight 359.19
Percent Purity 95%
Chemical Name or Material Tri-n-butyl(3-methyl-2-butenyl)tin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.