missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyl(vinyl)tin, 96%

Product Code. 15499129
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499129 25 g 25g
11329066 1 g 1g
11339066 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15499129 Supplier Thermo Scientific Alfa Aesar Supplier No. L20215.14
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7486-35-3 | C14H30Sn | 317.104 g/mol

Tributyl(vinyl)tin is used as a reagent in the palladium catalyzed synthesis of allyl and benzyl ethers. It is also a vinyl nucleophile for bromoacetylene and bromoaromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tributyl(vinyl)tin is used as a reagent in the palladium catalyzed synthesis of allyl and benzyl ethers. It is also a vinyl nucleophile for bromoacetylene and bromoaromatics.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7486-35-3
Molecular Formula C14H30Sn
MDL Number MFCD00009421
InChI Key QIWRFOJWQSSRJZ-UHFFFAOYSA-N
Synonym tributyl vinyl tin, vinyltributyltin, vinyltributylstannane, tributyl vinyl stannane, tributyl ethenyl stannane, stannane, tributylethenyl, vinyltributyl tin, stannane, tributylvinyl, tributylstannylethylene, vinyltri-n-butyltin
PubChem CID 81998
IUPAC Name tributyl(ethenyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C=C

Specifications

Density 1.085
Boiling Point 104°C to 106°C (3.5 mmHg)
Flash Point >110°C
Refractive Index 1.478
Quantity 25 g
UN Number UN2788
Beilstein 3537662
Sensitivity Moisture sensitive
Solubility Information Not miscible or difficult to mix aq. soln..
Molecular Weight (g/mol) 317.10
Formula Weight 317.11
Percent Purity 96%
Chemical Name or Material Tri-n-butyl(vinyl)tin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.