missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyltin hydride, 97%

Product Code. 11450883
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
250 g
Unit Size:
100g
10g
250g
25g
50g
Product Code. Quantity unitSize
11450883 100 g 100g
11453900 10 g 10g
11440883 25 g 25g
11463900 50 g 50g
11473900 250 g 250g
5 options
This item is not returnable. View return policy

Product Code. 11450883

Brand: Thermo Scientific Alfa Aesar A13298.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 688-73-3 | C12H27Sn | 290.058 g/mol

Tri-n-butyltin hydride is used as a radical reagent in reductive cleavage, radical dehalogenation and intramolecular radical cyclization. It also promotes intramolecular cyclization leading to isoxazolo-benzaulene ring system. Further, it acts as a reducing agent for the conversion of alkyl halides to hydrocarbons. In addition to this, it is used in a veterinary anthelmintic for poultry and an intermediate to make other butyltin compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butyltin hydride is used as a radical reagent in reductive cleavage, radical dehalogenation and intramolecular radical cyclization. It also promotes intramolecular cyclization leading to isoxazolo-benzaulene ring system. Further, it acts as a reducing agent for the conversion of alkyl halides to hydrocarbons. In addition to this, it is used in a veterinary anthelmintic for poultry and an intermediate to make other butyltin compounds.

Solubility
Immiscible with water.

Notes
Air and moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 688-73-3
Molecular Formula C12H27Sn
Molecular Weight (g/mol) 290.058
MDL Number MFCD00009416
InChI Key PIILXFBHQILWPS-UHFFFAOYSA-N
Synonym tributylstannanyl, tributlytin, tributylstannic hydride, tributyl tin, tri-n-butylstannane hydride, unii-4xdx163p3d, tin, tri-n-butyl-, hydride, tri-n-butylstannyl radical, stannane, tri-n-butyl-, hydride, tributyl-tin
PubChem CID 3032732
IUPAC Name tributyltin
SMILES CCCC[Sn](CCCC)CCCC

Specifications

Melting Point 0°C to 1°C
Density 1.098
Boiling Point 68°C to 74°C (0.3 mmHg)
Flash Point 40°C (104°F)
Odor Odorless
Refractive Index 1.473
Quantity 100 g
UN Number UN2929
Beilstein 3587329
Sensitivity Air sensitive; May form a precipitate
Solubility Information Immiscible with water.
Formula Weight 291.07
Percent Purity 97%
Chemical Name or Material Tri-n-butyltin hydride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.