missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyltin hydride, 97%

Product Code. 11463900
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
250 g
Unit Size:
100g
10g
250g
25g
50g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463900 50 g 50g
11453900 10 g 10g
11440883 25 g 25g
11450883 100 g 100g
11473900 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11463900 Supplier Thermo Scientific Alfa Aesar Supplier No. A13298.18
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 688-73-3 | C12H27Sn | 290.058 g/mol

Tri-n-butyltin hydride is used as a radical reagent in reductive cleavage, radical dehalogenation and intramolecular radical cyclization. It also promotes intramolecular cyclization leading to isoxazolo-benzaulene ring system. Further, it acts as a reducing agent for the conversion of alkyl halides to hydrocarbons. In addition to this, it is used in a veterinary anthelmintic for poultry and an intermediate to make other butyltin compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butyltin hydride is used as a radical reagent in reductive cleavage, radical dehalogenation and intramolecular radical cyclization. It also promotes intramolecular cyclization leading to isoxazolo-benzaulene ring system. Further, it acts as a reducing agent for the conversion of alkyl halides to hydrocarbons. In addition to this, it is used in a veterinary anthelmintic for poultry and an intermediate to make other butyltin compounds.

Solubility
Immiscible with water.

Notes
Air and moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 688-73-3
Molecular Formula C12H27Sn
Molecular Weight (g/mol) 290.06
MDL Number MFCD00009416
InChI Key PIILXFBHQILWPS-UHFFFAOYSA-N
Synonym tributylstannanyl, tributlytin, tributylstannic hydride, tributyl tin, tri-n-butylstannane hydride, unii-4xdx163p3d, tin, tri-n-butyl-, hydride, tri-n-butylstannyl radical, stannane, tri-n-butyl-, hydride, tributyl-tin
PubChem CID 3032732
IUPAC Name tributylstannyl
SMILES CCCC[Sn](CCCC)CCCC

Specifications

Melting Point 0°C to 1°C
Density 1.098
Boiling Point 68°C to 74°C (0.3 mmHg)
Flash Point 40°C (104°F)
Odor Odorless
Refractive Index 1.473
Quantity 50 g
UN Number UN2929
Beilstein 3587329
Sensitivity Air sensitive; May form a precipitate
Solubility Information Immiscible with water.
Formula Weight 291.07
Percent Purity 97%
Chemical Name or Material Tri-n-butyltin hydride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tri-n-butyltin hydride, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contain: Tri-n-butyltin hydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.