missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-octylphosphine, tech. 90%

Product Code. 15419029
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15419029 500 mL 500mL
11332767 25 mL 25mL
11342767 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15419029 Supplier Thermo Scientific Alfa Aesar Supplier No. L15744.AP
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4731-53-7 | C24H51P | 370.646 g/mol

Tri-n-octylphosphine is used for coating zinc sulfide shells on cadmium-selenium quantum dot core by successive ionic layer adsorption and reaction method. It acts as a precursor to trioctylphosphine oxide. Further, it is used as a solvent for cadmium and selenium precursors. It also serves as a common reagent in the chemical synthesis of nanoparticles. It acts as a precursor to trioctylphosphine oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-octylphosphine is used for coating zinc sulfide shells on cadmium-selenium quantum dot core by successive ionic layer adsorption and reaction method. It acts as a precursor to trioctylphosphine oxide. Further, it is used as a solvent for cadmium and selenium precursors. It also serves as a common reagent in the chemical synthesis of nanoparticles. It acts as a precursor to trioctylphosphine oxide.

Solubility
Immiscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4731-53-7
Molecular Formula C24H51P
Molecular Weight (g/mol) 370.646
MDL Number MFCD00015298
InChI Key RMZAYIKUYWXQPB-UHFFFAOYSA-N
Synonym trioctylphosphine, tri-n-octylphosphine, phosphine, trioctyl, tri-n-octyl phosphine, tri-octylphosphine, trioctyl phosphine, tri degrees ctylphosphine, acmc-209k8q
PubChem CID 20851
IUPAC Name trioctylphosphane
SMILES CCCCCCCCP(CCCCCCCC)CCCCCCCC

Specifications

Density 0.831
Boiling Point 240°C (10 mmHg)
Flash Point 147°C (296°F)
Odor Garlic-like
Refractive Index 1.468
Quantity 500 mL
UN Number UN1760
Beilstein 1776995
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Formula Weight 370.65
Percent Purity 90%
Grade Technical
Chemical Name or Material Tri-n-octylphosphine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.