missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-propyl orthoformate, 97%

Product Code. 11432897
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11432897

Brand: Thermo Scientific Alfa Aesar A15503.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 621-76-1 | C10H22O3 | 190.283 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-76-1
Molecular Formula C10H22O3
Molecular Weight (g/mol) 190.283
MDL Number MFCD00015214
InChI Key RWNXXQFJBALKAX-UHFFFAOYSA-N
Synonym tripropyl orthoformate, tri-n-propyl orthoformate, orthoformic acid tripropyl ester, 1-dipropoxymethoxy propane, unii-z8f0df6af9, propane, 1,1',1-methylidynetris oxy tris, z8f0df6af9, tripropoxymethane, tripropylorthoformate
PubChem CID 69311
IUPAC Name 1-(dipropoxymethoxy)propane
SMILES CCCOC(OCCC)OCCC

Specifications

Density 0.886
Boiling Point 106°C to 108°C (40 mmHg)
Flash Point 72°C (161°F)
Refractive Index 1.407
Quantity 5 g
Beilstein 1744249
Sensitivity Moisture sensitive
Formula Weight 190.28
Percent Purity 97%
Chemical Name or Material Tri-n-propyl orthoformate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.