missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-propylsilane, 99%, Thermo Scientific™

Product Code. 10697084 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10697084 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10697084 Supplier Thermo Scientific Acros Supplier No. 390320050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 998-29-8
Molecular Formula C9H22Si
Molecular Weight (g/mol) 158.36
MDL Number MFCD00009358
InChI Key MMYRBBZVCDXGHG-UHFFFAOYSA-N
Synonym tripropylsilane, tri-n-propylsilane, silane, tripropyl, silicon tri-n-propyl, tripropyl silane, tri-n-propyl silane, zhovawfvvbwegq-uhfffaoysa-n
PubChem CID 6327709
IUPAC Name tripropylsilicon
SMILES CCC[Si](CCC)CCC

Specifications

Color Colorless
Density 0.7580g/mL
Boiling Point 173°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HSi(CH2CH2CH3)3
Refractive Index 1.4270 to 1.429
Quantity 5 g
Specific Gravity 0.758
Solubility Information Solubility in water: insoluble
Formula Weight 158.36
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tri-n-propylsilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tri-n-propylsilane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.