missing translation for 'onlineSavingsMsg'
Learn More

tri-n-Propyltin Chloride, TRC

Product Code. 30414594
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30414594 250 mg
30415808 1 g
30416592 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30414594 Supplier Toronto Research Chemicals Supplier No. TRCP838825250MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2279-76-7
Molecular Formula C9 H21 Cl Sn
Molecular Weight (g/mol) 283.43
Synonym Tripropyltin chloride, TPTCl
IUPAC Name chloro(tripropyl)stannane
SMILES CCC[Sn](Cl)(CCC)CCC

Specifications

Quantity 250 mg
InChI Formula InChI=1S/3C3H7.ClH.Sn/c3*1-3-2;;/h3*1,3H2,2H3;1H;/q;;;;+1/p-1
Formula Weight 284.0354
Chemical Name or Material Tri-n-propyltin chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.