missing translation for 'onlineSavingsMsg'
Learn More

Tri-O-acetyl-D-glucal, TRC

Product Code. 30452392
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452392 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30452392 Supplier Toronto Research Chemicals Supplier No. TRCT76480025G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tri-O-acetyl-D-glucal
CAS 2873-29-2
Molecular Formula C12 H16 O7
Quantity 25 g
Synonym D-arabino-Hex-1-enitol','1,5-anhydro-2-deoxy-','3,4,6-triacetate, D-arabino-Hex-1-enitol','1,5-anhydro-2-deoxy-','triacetate (9CI), D-arabino-Hex-1-enopyranose','1,2-dideoxy-','triacetate (8CI), 3,4,6-Tri-O-acetyl-1,2-dideoxy-D-arabino-hex-1-enopyranose, 3,4,6-Tri-O-acetyl-D-glucal, 3,4,6-Tri-O-acetylglucal, 3,4,6-Tris-O-acetyl-D-glucal, D-Glucal triacetate, Tri-O-acetyl-D-glucal, Tri-O-acetylglucal, Triacetyl-D-glucal
InChI Formula InChI=1S/C12H16O7/c1-7(13)17-6-11-12(19-9(3)15)10(4-5-16-11)18-8(2)14/h4-5,10-12H,6H2,1-3H3/t10-,11-,12+/m1/s1
SMILES CC(=O)OC[C@H]1OC=C[C@@H](OC(=O)C)[C@@H]1OC(=O)C
IUPAC Name [(2R,3S,4R)-3,4-diacetyloxy-3,4-dihydro-2H-pyran-2-yl]methyl acetate
Molecular Weight (g/mol) 272.2512
Formula Weight 272.0896
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.