missing translation for 'onlineSavingsMsg'
Learn More

Tri-o-tolyl phosphate, 96%

Product Code. 10015403 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10015403 25 g Glass bottle 25g
10613193 100 g Glass bottle 100g
10324312 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10015403 Supplier Thermo Scientific Acros Supplier No. 366460250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-30-8 | C21H21O4P | 368.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-30-8
Molecular Formula C21H21O4P
Molecular Weight (g/mol) 368.37
MDL Number MFCD00041906
InChI Key YSMRWXYRXBRSND-UHFFFAOYSA-N
Synonym tri-o-cresyl phosphate, tri-o-tolyl phosphate, o-tolyl phosphate, tocp, totp, phosphoric acid, tris 2-methylphenyl ester, o-cresyl phosphate, triorthocresyl phosphate, phosflex 179c, tri-2-tolyl phosphate
PubChem CID 6527
IUPAC Name tris(2-methylphenyl) phosphate
SMILES CC1=CC=CC=C1OP(=O)(OC1=CC=CC=C1C)OC1=CC=CC=C1C

Specifications

Melting Point -25°C
Color Colorless to Yellow
Density 1.2000g/mL
Boiling Point 410°C
Flash Point 225°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5580 to 1.561
Quantity 25 g
Merck Index 15, 9941
Specific Gravity 1.2
Solubility Information Solubility in water: 0.01g/100mL (25°C). Other solubilities: slightly soluble in alcohol,soluble in ether
Formula Weight 368.36
Percent Purity 96%
Physical Form Viscous Liquid
Chemical Name or Material Tri-o-tolyl phosphate, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tri-o-tolyl phosphate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H370-Causes damage to organs.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.