missing translation for 'onlineSavingsMsg'
Learn More

Tri-Potassium Citrate, Extra Pure, SLR, Fisher Chemical

Product Code. 10785721 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10785721 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10785721 Supplier Fisher Chemical Supplier No. P/4440/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 180°C
CAS 6100-05-6
Color White
pH 7.5 to 9.0
Physical Form Solid
Quantity 500 g
Molecular Formula C6H7K3O8
MDL Number 150442
Synonym potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
InChI Key PJAHUDTUZRZBKM-UHFFFAOYSA-K
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
IUPAC Name tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
Molecular Weight (g/mol) 324.41
PubChem CID 2735208
ChEBI CHEBI:64746
Formula Weight 324.42g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.