missing translation for 'onlineSavingsMsg'
Learn More

Tri-Sodium Citrate Dihydrate, HPLC for Electrochemical Detection, Certified, Fisher Chemical

Product Code. 10030520 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10030520

Brand: Fisher Chemical S/3380/53

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point >300°C
CAS 4-3-6132
Color White
pH 8.4
Physical Form Solid
Quantity 500 g
Molecular Formula C6H9Na3O9
MDL Number 150031
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
Molecular Weight (g/mol) 294.10
PubChem CID 71474
ChEBI CHEBI:32142
Formula Weight 294.09g/mol
Packaging Plastic powder jar
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.