missing translation for 'onlineSavingsMsg'
Learn More

Tri-tert-butyl borate, 97%

Product Code. 15451708
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15451708 5 g 5g
15461708 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15451708 Supplier Thermo Scientific Alfa Aesar Supplier No. H55461.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7397-43-5 | C12H27BO3 | 230.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7397-43-5
Molecular Formula C12H27BO3
Molecular Weight (g/mol) 230.155
MDL Number MFCD00009652
InChI Key ZMCWFMOZBTXGKI-UHFFFAOYSA-N
Synonym tri-tert-butyl borate, boron tert-butoxide, tri-tert-butoxyborane, tris tert-butyl borate, tris tert-butyloxy borane, boric acid h3bo3 , tri-tert-butyl ester, boric acid,tris 1,1-dimethylethyl ester, tri-t-butylborate, tri-t-butyl borate, tritert-butoxy-borane
PubChem CID 289737
IUPAC Name tritert-butyl borate
SMILES B(OC(C)(C)C)(OC(C)(C)C)OC(C)(C)C

Specifications

Melting Point 18°C to 19°C
Color Colorless
Density 0.811 g/mL
Boiling Point 101°C (74 mmHg)
Flash Point 29°C (84°F)
Linear Formula [(CH3)3CO]3B
Refractive Index 1.389
Quantity 5 g
UN Number UN1993
Formula Weight 230.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tri-tert-butyl borate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.