missing translation for 'onlineSavingsMsg'
Learn More

Triacetin, 99%

Product Code. 10275330 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10275330 500 mL Glass bottle 500mL
10215240 1 L Glass bottle 1 L
10021490 2.5 L Glass bottle 2.5 L
11974911 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10275330 Supplier Thermo Scientific Acros Supplier No. 139225000
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 102-76-1 | C9H14O6 | 218.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-76-1
Molecular Formula C9H14O6
Molecular Weight (g/mol) 218.21
InChI Key URAYPUMNDPQOKB-UHFFFAOYSA-N
Synonym triacetin, glyceryl triacetate, glycerol triacetate, glycerin triacetate, enzactin, triacetine, triacetylglycerol, fungacetin, glyped, triacetyl glycerine
PubChem CID 5541
ChEBI CHEBI:9661
IUPAC Name 2,3-diacetyloxypropyl acetate
SMILES CC(=O)OCC(COC(=O)C)OC(=O)C

Specifications

Melting Point 3.0°C
Color Colorless
Density 1.1550g/mL
Boiling Point 258.0°C
Flash Point 138°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3CO2CH2)2CH(O2CCH3)
Refractive Index 1.4290 to 1.4330
Quantity 500 mL
Beilstein 02,147
Merck Index 15,9751
Specific Gravity 1.155
Solubility Information Solubility in water: 64.0g/L (20°C). Other solubilities: soluble in most common organic solvents,slightly soluble in carbon disulfide
Viscosity 23 mPa.s (20°C)
Formula Weight 218.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Triacetin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.