missing translation for 'onlineSavingsMsg'
Learn More

Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, 98%, Stabilized

Product Code. 10587231 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10587231 100 g 100g
10458742 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10587231 Supplier Thermo Scientific Acros Supplier No. 342121000
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1025-15-6 | C12H15N3O3 | 249.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1025-15-6
Molecular Formula C12H15N3O3
Molecular Weight (g/mol) 249.27
MDL Number MFCD00006554
InChI Key KOMNUTZXSVSERR-UHFFFAOYSA-N
Synonym triallyl isocyanurate, isocyanuric acid triallyl ester, 1,3,5-triallyl-1,3,5-triazinane-2,4,6-trione, triallylisocyanurate, taic, diak 7, 1,3,5-triallylisocyanurate, 1,3,5-triallyl isocyanurate, 1,3,5-triallylisocyanuric acid, 1,3,5-tri-2-propenyl-1,3,5-triazine-2,4,6 1h,3h,5h-trione
PubChem CID 13931
IUPAC Name 1,3,5-tris(prop-2-enyl)-1,3,5-triazinane-2,4,6-trione
SMILES C=CCN1C(=O)N(C(=O)N(C1=O)CC=C)CC=C

Specifications

Melting Point 24.8°C
Color Colorless to Yellow
Density 1.1590g/mL
Boiling Point 149.0°C to 152.0°C (4.0 mmHg)
Flash Point 245°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5120 to 1.5140
Quantity 100 g
Beilstein 26, IV, 637
Specific Gravity 1.159
Solubility Information Solubility in water: >1g/L (20°C). Other solubilities: soluble in acetone,dimethyl sulfoxide,ethanol
Stabilizer 1200ppm MEHQ
Viscosity 90.1 mPa.s (30°C)
Formula Weight 249.27
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.