missing translation for 'onlineSavingsMsg'
Learn More

Triamterene, TRC

Product Code. 30132621
Change view
Click to view available options
Quantity:
50 mg
5 g
10 g
Unit Size:
10g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132621 10 g 10g
30003798 50 mg 50mg
30011945 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132621 Supplier Toronto Research Chemicals Supplier No. TRCT76732510G
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 396-01-0
Molecular Formula C12 H11 N7
Molecular Weight (g/mol) 253.26
Synonym 6-Phenyl-2,4,7-pteridinetriamine, 2,4,7-Triamino-6-phenyl-pteridine, 2,4,7-Triamino-6-phenylpteridine, 6-Phenyl-2,4,7-triaminopteridine, Ademin, Ademine, Diren, Ditak, Diurene, Dyren, Dyrenium, Dytac, Jatropur, NSC 77625, Noridil, Pterofen, Pterophene, SKF 8542, Taturil, Teriam, Teridin, Tri-Span, Triampur, Triamteren, Triamterene, Triamteril, Triamteril complex, Triteren, Urocaudal
IUPAC Name 6-phenylpteridine-2,4,7-triamine
SMILES Nc1nc(N)c2nc(c(N)nc2n1)c3ccccc3

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H11N7/c13-9-7(6-4-2-1-3-5-6)16-8-10(14)18-12(15)19-11(8)17-9/h1-5H,(H6,13,14,15,17,18,19)
Formula Weight 253.1076
Chemical Name or Material Triamterene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.