missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Triangular Tripod Stand

Product Code. 12966916 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
125 mm side length
150 mm side length
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12966916 150 mm side length Each
11779806 125 mm side length Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12966916
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Designed to cope with the demands of a modern laboratory.

Zinc plated steel with 210mm height, 6mm diameter legs.

Specifications

Material Stainless Steel
Height (Metric) 210 mm
Length (Metric) 150 mm side length
Product Type Tripod Stand
Shape Triangular
Finish Zinc Plated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.