missing translation for 'onlineSavingsMsg'
Learn More

Tribenzylphosphine

Product Code. 11331228
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11331228 25 g 25g
11321228 1 g 1g
11341228 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11331228 Supplier Thermo Scientific Alfa Aesar Supplier No. 010173.14
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7650-89-7 | C21H21P | 304.37 g/mol

Tribenzylphosphine is used as a co catalyst in preparation of coumarin derivatives via nickel-catalyzed cycloaddition, copolymerization reactions, Rh-catalyzed hydroformylation reactions, preparation of substituted allylamines by three-component coupling of alkynes, imines and organoboranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7650-89-7
Molecular Formula C21H21P
Molecular Weight (g/mol) 304.37
MDL Number MFCD00014439
InChI Key IFXORIIYQORRMJ-UHFFFAOYSA-N
Synonym tribenzylphosphine, phosphine, tris phenylmethyl, tribenzyl phosphine, tris phenylmethyl phosphane, tris phenylmethyl phosphine, phosphine, tribenzyl, acmc-20ajfj, tribenzylphosphine 1g
PubChem CID 82101
IUPAC Name tribenzylphosphane
SMILES C(P(CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 93°C to 99°C
Color White
Boiling Point 183°C to 185°C (0.2 mmHg)
Linear Formula (C6H5CH2)3P
Quantity 25 g
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 304.37
Physical Form Crystalline
Chemical Name or Material Tribenzylphosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.