missing translation for 'onlineSavingsMsg'
Learn More

Tributyl(1-ethoxyvinyl)tin, 95%

Product Code. 10557513 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10557513 5 g Glass bottle 5g
10413063 1 g Glass bottle 1g
10338893 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10557513 Supplier Thermo Scientific Acros Supplier No. 377710050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 97674-02-7 | C16H34OSn | 361.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 97674-02-7
Molecular Formula C16H34OSn
Molecular Weight (g/mol) 361.16
MDL Number MFCD00010240
InChI Key HGXJOXHYPGNVNK-UHFFFAOYSA-N
Synonym tributyl 1-ethoxyvinyl stannane, tributyl 1-ethoxyvinyl tin, tributyl 1-ethoxyethenyl stannane, 1-ethoxyvinyltributyltin, 1-ethoxyvinyl tributyltin, 1-ethoxyvinyltri-n-butyltin, 1-ethoxyvinyl tributylstannane, 1-ethoxyethenyl tributylstannane, tributyl-1-ethoxyvinyl tin, stannane, tributyl 1-ethoxyethenyl
PubChem CID 619414
IUPAC Name tributyl(1-ethoxyethenyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C(=C)OCC

Spezifikation

Color Colorless
Density 1.0690g/mL
Boiling Point 85.0°C to 86.0°C (0.1 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula [CH3(CH2)3]SnC(CH2)OCH2CH3
Refractive Index 1.4750 to 1.4770
Quantity 5 g
Specific Gravity 1.069
Solubility Information Solubility in water: insoluble
Formula Weight 361.16
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tributyl(1-ethoxyvinyl)tin
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Tributyl(1-ethoxyvinyl)tin
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Gefahrenhinweis
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Sicherheitshinweis
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.