missing translation for 'onlineSavingsMsg'
Learn More

Tributyl(3-methyl-2-butenyl)tin, 90%, ACROS Organics™

Product Code. 10282432 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10282432 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10282432 Supplier Thermo Scientific Acros Supplier No. 370220010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 53911-92-5
Color Colorless
Density 1.0690g/mL
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Molecular Formula C17H36Sn
Linear Formula [CH3(CH2)3]3SnCH2CH=C(CH3)2
Refractive Index 1.4860 to 1.4900 (20°C, 589nm)
MDL Number MFCD01863649
Quantity 1 g
Synonym tributyl 3-methylbut-2-en-1-yl stannane, tributyl 3-methyl-2-butenyl tin, tributylprenylstannane, acmc-1am9p, tri-n-butyl 3-methyl-2-butenyl tin, tributyl 3-methylbut-2-enyl stannane, stannane, tributyl 3-methyl-2-butenyl, tributyl 3-methylbut-2-en-1-yl ;stannane
InChI Key XEFRYQPTIWSVGI-UHFFFAOYSA-N
SMILES CCCC[Sn](CCCC)(CCCC)CC=C(C)C
IUPAC Name tributyl(3-methylbut-2-enyl)stannane
Molecular Weight (g/mol) 359.185
PubChem CID 2734722
Formula Weight 359.18
Percent Purity ≥88%
Physical Form Liquid
Chemical Name or Material Tributyl(3-methyl-2-butenyl)tin, 90%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tributyl(3-methyl-2-butenyl)tin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.