missing translation for 'onlineSavingsMsg'
Learn More

Tributyl borate, 98%, AcroSeal™

Product Code. 10606240 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10606240 100 mL AcroSeal™ Glass Bottle 100mL
10566230 800 mL AcroSeal™ Glass Bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10606240 Supplier Thermo Scientific Acros Supplier No. 428661000
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 688-74-4 | C12H27BO3 | 230.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 688-74-4
Molecular Formula C12H27BO3
Molecular Weight (g/mol) 230.15
MDL Number MFCD00009434
InChI Key LGQXXHMEBUOXRP-UHFFFAOYSA-N
Synonym n-butyl borate, tri-n-butyl borate, tributoxyborane, butyl borate, tributoxyboron, boron tributoxide, borester 2, tri-n-butoxyborane, tris butoxy borane, borane, tributoxy
PubChem CID 12712
IUPAC Name tributyl borate
SMILES B(OCCCC)(OCCCC)OCCCC

Specifications

Melting Point -70.0°C
Color Colorless
Density 0.8530g/mL
Boiling Point 230.0°C to 235.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula (CH3CH2CH2CH2O)3B
Refractive Index 1.4080 to 1.4100
Quantity 100 mL
Specific Gravity 0.853
Solubility Information Solubility in water: decomposes. Other solubilities: miscible with most common organic liquids
Formula Weight 230.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tributyl borate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.