missing translation for 'onlineSavingsMsg'
Learn More

Tributyl Phosphate, BAKER™, J.T. Baker™

Product Code. 15588834 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15588834 500 mL Narrow Mouth Amber Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15588834 Supplier Avantor J.T.Baker Supplier No. W43207
  • ONLINE EXCLUSIVE PRICE
    107.57 €
    86.68 € / 500mL
    Save 20.89 €
    19% Off
 Restricted Item
Estimated Shipment: 11-06-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A grade of chemicals for laboratory use

Specifications

CAS 126-73-8
Flash Point 120.00°C
Packaging Narrow Mouth Amber Glass Bottle
Molecular Formula C12H27O4P
MDL Number MFCD00009436
Quantity 500 mL
Specific Gravity 1L = 0.98kg
Synonym tri-n-butyl phosphate, tributylphosphate, butyl phosphate, phosphoric acid tributyl ester, tributylphosphat, celluphos 4, disflamoll tb, tributilfosfato, tributylfosfaat, tributyle phosphate de
InChI Key STCOOQWBFONSKY-UHFFFAOYSA-N
SMILES CCCCOP(=O)(OCCCC)OCCCC
IUPAC Name tributyl phosphate
Molecular Weight (g/mol) 266.32
PubChem CID 31357
ChEBI CHEBI:35019
Formula Weight 266.32
Chemical Name or Material Tributyl Phosphate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tributyl phosphate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Tributyl phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.