missing translation for 'onlineSavingsMsg'
Learn More

Tributyl phosphite, 94%, ACROS Organics™

Product Code. 10617532 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1kg; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10617532 1kg; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10617532 Supplier Thermo Scientific Acros Supplier No. 169920010
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 102-85-2, 71-36-3
Color Colorless
Density 0.915g/mL
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Molecular Formula C12H27O3P
Linear Formula {CH3(CH2)3O}3P
Refractive Index 1.4310 to 1.4330 (20°C, 589nm)
MDL Number MFCD00009437
Quantity 1 kg
Beilstein 01, I, 187
Fieser 15,280
Synonym tri-n-butyl phosphite, phosphorous acid, tributyl ester, tributoxyphosphine, tributylfosfit, butyl phosphite c4h9o 3p, tributylfosfit czech, tributylphosphite, phosphorus tributoxide p obu 3, tri-n-butylphosphite, 1-butanol, 1,1',1-phosphinidynetri
InChI Key XTTGYFREQJCEML-UHFFFAOYSA-N
SMILES CCCCOP(OCCCC)OCCCC
IUPAC Name tributyl phosphite
Molecular Weight (g/mol) 250.319
PubChem CID 7623
Formula Weight 250.32
Percent Purity ≥92%
Physical Form Liquid
Chemical Name or Material Tributyl phosphite, 94%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.