missing translation for 'onlineSavingsMsg'
Learn More

Tributylphenyltin

Product Code. 10661853 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Packungsgröße:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packaging Packungsgröße
10661853 10 g Glass bottle 10g
10384792 1 g Glass bottle 1g
10156943 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 10661853 Lieferant Thermo Scientific Acros Lieferanten-Nr. 370230100
 Restricted Item
Estimated Shipment: 03-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 960-16-7 | C18H32Sn | 367.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 960-16-7
Molecular Formula C18H32Sn
Molecular Weight (g/mol) 367.16
MDL Number MFCD00134394
InChI Key SYUVAXDZVWPKSI-UHFFFAOYSA-N
Synonym tributyl phenyl stannane, tributylphenyltin, phenyltributylstannane, stannane, tributylphenyl, phenyltributyltin, phenyltri-n-butyltin, phenyltributlytin, tributylphenyl tin, phenyl tnbutyl tin, phenyl tributyl tin
PubChem CID 607632
IUPAC Name tributyl(phenyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CC=C1

Spezifikation

Color Colorless
Density 1.1250g/mL
Boiling Point 125.0°C to 128.0°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5Sn[(CH2)3CH3]3
Refractive Index 1.5150 to 1.5170
Quantity 10 g
Specific Gravity 1.125
Formula Weight 367.16
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Tributylphenyltin
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Tributylphenyltin
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Gefahrenhinweis
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360FD-May damage fertility. May damage the unborn child.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Sicherheitshinweis
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.