missing translation for 'onlineSavingsMsg'
Learn More

Tributyrin, 97%

Product Code. 10554264 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
250mL
This item is not returnable. View return policy

Product Code. 10554264

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tributyrin, 97%, C15H26O6, CAS Number-60-01-5, butyrin, butyryl triglyceride, glycerol tributanoate, glycerol tributyrate, glyceroltributyrin, glyceryl tributyrate, propane-1,2,3-triyl tributyrate, tributin, tributyrin, tri-n-butyrin, 100mL, Liquid, 96% min. (GC), 02, 273, 305.0 deg.C to 310.0 deg.C | CAS: 60-01-5 | C15H26O6 | 302.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-01-5
Molecular Formula C15H26O6
Molecular Weight (g/mol) 302.36
MDL Number MFCD00009392
InChI Key UYXTWWCETRIEDR-UHFFFAOYSA-N
Synonym tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin
PubChem CID 6050
ChEBI CHEBI:35020
IUPAC Name 2,3-di(butanoyloxy)propyl butanoate
SMILES CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC

Specifications

Melting Point -75.0°C
Color Clear colorless to light yellow
Density 1.0335g/mL
Boiling Point 305.0°C to 310.0°C
Flash Point 173°C
Infrared Spectrum Conforms
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3CH2CH2CO2CH2)2CH(O2CCH2CH2CH3)
Refractive Index 1.4340 to 1.437
Quantity 1 L
Beilstein 02, 273
Merck Index 15, 9785
Specific Gravity 1.0335
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 302.36
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Tributyrin, 98%
Mostrar más Mostrar menos

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado