missing translation for 'onlineSavingsMsg'
Learn More

Tributyrin, 98.4%, MP Biomedicals™

Product Code. 11403511 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11403511 100 mL 100mL
11413511 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11403511 Supplier MP Biomedicals Supplier No. 0210311180
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 60-01-5
Molecular Formula C15H26O6
InChI Key UYXTWWCETRIEDR-UHFFFAOYSA-N
SMILES CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC

Specifications

Color Colorless
Density 1.032g/mL
Assay Percent Range 0.984
Quantity 100 mL
Synonym tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin
IUPAC Name 2,3-di(butanoyloxy)propyl butanoate
Molecular Weight (g/mol) 302.367
PubChem CID 6050
ChEBI CHEBI:35020
Formula Weight 302.36
Physical Form Liquid
Chemical Name or Material Tributyrin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.