missing translation for 'onlineSavingsMsg'
Learn More

Tricarbonyldichlororuthenium(II) dimer

Product Code. 15470987
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470987 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15470987 Supplier Thermo Scientific Alfa Aesar Supplier No. 010503.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 22594-69-0 | C6Cl4O6Ru2 | 512.00 g/mol

Tricarbonyldichlororuthenium(II) dimer is used to enhance the coagulation and attenuation of vulnerability to fibrinolysis, modify thrombus growth or disintegration, enhances fibrinogen as a substrate for thrombin, and regulation of ion transport by gasotransmitters. Also used for various pharmacological studies. It is used as a CO donor for reactive oxygen species mediated bacterial killing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 22594-69-0
Molecular Formula C6Cl4O6Ru2
MDL Number MFCD00011528
Quantity 1 g
UN Number UN3466
Synonym corm-2, tricarbonyldichlororuthenium ii dimer, rucl2 co 3 2, dichlorotricarbonylruthenium ii dimer, carbon monooxide-dichlororuthenium 3/1
Solubility Information Insoluble in water
InChI Key JYHHJVKGDCZCCL-UHFFFAOYSA-J
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Ru++].[Ru++].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
IUPAC Name carbon monoxide;dichlororuthenium
Molecular Weight (g/mol) 512.00
PubChem CID 10951331
Formula Weight 512.02
Physical Form Crystalline
Chemical Name or Material Tricarbonyldichlororuthenium(II) dimer
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.