missing translation for 'onlineSavingsMsg'
Learn More

Tricarbonylnitrosylcobalt

Product Code. 11312628
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312628 1 g 1g
11302628 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312628 Supplier Thermo Scientific Alfa Aesar Supplier No. 023151.03
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14096-82-3

Tricarbonylnitrosylcobalt is used as the Co metalorganic precursor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14096-82-3
MDL Number MFCD00016014
Synonym cobalt tricarbonyl nitrosyl, tricarbonyl-nitrosyl-cobalt

Specifications

Melting Point -1°C
Color Red
Density 1.47 g/mL
Boiling Point 48.6°C (55°C (decomposition))
Odor Unpleasant
Linear Formula Co(NO)(CO)3
Quantity 1 g
UN Number UN1992
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 172.97
Physical Form Liquid
Chemical Name or Material Tricarbonylnitrosylcobalt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.