missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic anhydride, 95%

Product Code. 12695017 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12695017 25 g 25g
12605027 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12695017 Supplier Thermo Scientific Acros Supplier No. 446090250
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4124-31-6 | C4Cl6O3 | 308.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4124-31-6
Molecular Formula C4Cl6O3
Molecular Weight (g/mol) 308.76
InChI Key MEFKFJOEVLUFAY-UHFFFAOYSA-N
Synonym trichloroacetic anhydride, acetic acid, trichloro-, anhydride, acetic acid, 2,2,2-trichloro-, 1,1'-anhydride, acmc-1aliy, perchloroacetic anhydride, bis trichloroacetic anhydride, trichloroacetic acid anhydride, 2,2,2-trichloroacetic anhydride, trichloroacetyl 2,2,2-trichloroacetate
PubChem CID 20079
IUPAC Name (2,2,2-trichloroacetyl) 2,2,2-trichloroacetate
SMILES C(=O)(C(Cl)(Cl)Cl)OC(=O)C(Cl)(Cl)Cl

Specifications

Density 1.6900g/mL
Boiling Point 139°C to 141°C (60.0 mmHg)
Packaging Glass Bottle
Linear Formula [CCl3C(=O)]2O
Quantity 25 g
Specific Gravity 1.69
Solubility Information Solubility in water: reacts
Formula Weight 308.76
Percent Purity 95%
Chemical Name or Material Trichloroacetic anhydride
compliance-icons
Product Identifier
  • Trichloroacetic anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.