missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroacetic Anhydride, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Trichloroacetic Anhydride |
| CAS | 4124-31-6 |
| Molecular Formula | C4Cl6O3 |
| Quantity | 5 g |
| Synonym | 2,2,2-Trichloro-acetic Acid 1,1'-Anhydride, Trichloracetic Anhydride, Trichloroacetic acid anhydride |
| InChI Formula | InChI=1S/C4Cl6O3/c5-3(6,7)1(11)13-2(12)4(8,9)10 |
| SMILES | ClC(Cl)(Cl)C(OC(C(Cl)(Cl)Cl)=O)=O |
| IUPAC Name | 2,2,2-trichloroacetic anhydride |
| Molecular Weight (g/mol) | 308.76 |
| Formula Weight | 305.8 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction