missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetonitrile, 98%

Product Code. 11681419 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11681419 2.5 kg Glass bottle 2.5kg
10604812 5 g Glass bottle 5g
10437530 100 g Glass bottle 100g
10274710 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11681419 Supplier Thermo Scientific Supplier No. 139370025
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 545-06-2 | C2Cl3N | 144.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 545-06-2
Molecular Formula C2Cl3N
Molecular Weight (g/mol) 144.39
InChI Key DRUIESSIVFYOMK-UHFFFAOYSA-N
Synonym trichloroacetonitrile, acetonitrile, trichloro, tritox, cyanotrichloromethane, trichloromethyl cyanide, trichloroethanenitrile, trichlouracetonitril, trichlor-acetonitrile, trichloromethylnitrile, nitrile trichloracetique
PubChem CID 11011
ChEBI CHEBI:82541
IUPAC Name 2,2,2-trichloroacetonitrile
SMILES C(#N)C(Cl)(Cl)Cl

Specifications

Melting Point -42.0°C
Color Colorless to Yellow
Density 1.4400g/mL
Boiling Point 83.0°C to 84.0°C
Flash Point 195°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CCl3CN
Refractive Index 1.4400 to 1.4420
Quantity 2.5 kg
Beilstein 02,212
Fieser 01,1194; 05,686; 06,604; 07,381; 12,526; 13,32
Merck Index 15,9793
Specific Gravity 1.44
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone,dimethyl sulfoxide,ethanol
Formula Weight 144.39
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Trichloroacetonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.