missing translation for 'onlineSavingsMsg'
Learn More

Trichloroethylene, Technical, Stabilised, Fisher Chemical

Product Code. 11428902 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber glass bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11428902 500 mL Amber glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11428902 Supplier Fisher Chemical Supplier No. T/3050/08
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

CAS 79-01-6
Melting Point -85°C
Color Colorless
Boiling Point 87°C
Packaging Amber glass bottle
Molecular Formula C2HCl3
MDL Number 838
Quantity 500 mL
Synonym trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
InChI Key XSTXAVWGXDQKEL-UHFFFAOYSA-N
SMILES C(=C(Cl)Cl)Cl
IUPAC Name 1,1,2-trichloroethene
Molecular Weight (g/mol) 131.38
PubChem CID 6575
Vapor Density 4.5
ChEBI CHEBI:16602
Formula Weight 131.39g/mol
Vapor Pressure 77.3mbar at 20°C
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trichloroethylene, stabilized
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H336-May cause drowsiness or dizziness.
  • H412-Harmful to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.