missing translation for 'onlineSavingsMsg'
Learn More

Trichloroisocyanuric acid, 99%

Product Code. 10711072 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10711072 250 g Glass Bottle 250g
10667581 1 kg Glass Bottle 1kg
11926581 5 kg Plastic bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10711072 Supplier Thermo Scientific Acros Supplier No. 421542500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 87-90-1 | C3Cl3N3O3 | 232.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-90-1
Molecular Formula C3Cl3N3O3
Molecular Weight (g/mol) 232.40
MDL Number MFCD00006553
InChI Key YRIZYWQGELRKNT-UHFFFAOYSA-N
Synonym trichloroisocyanuric acid, symclosene, trichlorocyanuric acid, symclosen, isocyanuric chloride, fichlor 91, trichloro-s-triazinetrione, trichloroisocyanic acid, fi clor 91, chloreal
PubChem CID 6909
ChEBI CHEBI:33015
SMILES ClN1C(=O)N(Cl)C(=O)N(Cl)C1=O

Specifications

Melting Point 245°C to 251°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Iodometry)
Packaging Glass Bottle
Quantity 250 g
Beilstein 26,256
Fieser 06,605; 13,322
Merck Index 15,9806
Solubility Information Solubility in water: 12g/L (25°C). Other solubilities: decomposes forming hclo,soluble in highly polar solvents
IUPAC Name 1,3,5-trichloro-1,3,5-triazinane-2,4,6-trione
Formula Weight 232.41
Percent Purity 99%
Physical Form Powder or Granules
Chemical Name or Material Trichloroisocyanuric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.