missing translation for 'onlineSavingsMsg'
Learn More

Trichloroisocyanuric Acid, TRC

Product Code. 30450349
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450349 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450349 Supplier Toronto Research Chemicals Supplier No. TRCT77399050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 87-90-1
Molecular Formula C3Cl3N3O3
Quantity 50 g
Synonym s-Triazine-2,4,6(1H,3H,5H)-trione, 1,3,5-trichloro- (8CI), s-Triazine-2,4,6(1H,3H,5H)-trione, trichloro- (6CI), 1,3,5-Trichloro-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, 1,3,5-Trichloro-1,3,5-triazinane-2,4,6-trione, 1,3,5-Trichloro-1,3,5-triazine-2,4,6-trione, 1,3,5-Trichloro-2,4,6-triazinetrione, 1,3,5-Trichloro-2,4,6-trioxohexahydro-s-triazine, 1,3,5-Trichloroisocyanuric acid, 1,3,5-Trichlorotriazine-2,4,6-trione, ACL 85, ACL 90, ACL 90 Plus, CDB 90, Chloreal, Doristrip W, Fi Clor 91, Hi-Lite 90, Hi-Lite 90G, Isocyanuric chloride, N,N',N' '-Trichloroisocyanuric acid, NSC 405124, Neochlor 90, Neochlor 90FG, Neochlor 90G, Superclean 90TH, Symclosen, Symclosene, TCCA, TICA-G, Trichloro-s-triazine-2,4,6(1H,3H,5H)-trione, Trichloro-s-triazinetrione, Trichlorocyanuric acid, Trichloroiminocyanuric acid, Trichloroisocyanuric acid, 1,3,5-Triazine-2,4,6(1H,3H,5H)-trione, 1,3,5-trichloro-, 1,3,5-Trichloro-s-triazine-2,4,6(1H,3H,5H)-trione, Trichloro-s-triazine-2,4,6(1H,3H,5H)-trione, 1,3,5-Trichloro-1,3,5-triazine-2,4,6-trione, 1,3,5-Trichloro-2,4,6-trioxohexahydro-s-triazine, 1,3,5-Trichloroisocyanuric acid, ACL 85, ACL 90, ACL 90 Plus, CDB 90, Chloreal, Doristrip W, Fi Clor 91, Hi-Lite 90, Hi-Lite 90G, Isocyanuric chloride, N,N',N' '-Trichloroisocyanuric acid, NSC 405124, Neochlor 90, Neochlor 90FG, Neochlor 90G, Superclean 90TH, Symclosen, Symclosene, TCCA, TCCA (chlorination agent), TICA-G, Trichloro-s-triazine-2,4,6(1H,3H,5H)-trione, Trichloro-s-triazinetrione, Trichlorocyanuric acid, Trichloroiminocyanuric acid, Trichloroisocyanuric acid
InChI Formula InChI=1S/C3Cl3N3O3/c4-7-1(10)8(5)3(12)9(6)2(7)11
SMILES ClN1C(=O)N(Cl)C(=O)N(Cl)C1=O
IUPAC Name 1,3,5-trichloro-1,3,5-triazinane-2,4,6-trione
Molecular Weight (g/mol) 232.41
Formula Weight 230.9005
Percent Purity >95%
Chemical Name or Material Trichloroisocyanuric Acid
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.