missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha,alpha-Trichlorotoluene, 98%

Product Code. 10551801 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10551801 5 mL Glass bottle 5mL
10397290 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10551801 Supplier Thermo Scientific Acros Supplier No. 164530050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 98-07-7 | C7H5Cl3 | 195.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-07-7
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.47
MDL Number MFCD00000786
InChI Key XEMRAKSQROQPBR-UHFFFAOYSA-N
Synonym benzotrichloride, trichloromethyl benzene, alpha,alpha,alpha-trichlorotoluene, phenylchloroform, toluene trichloride, benzenyl trichloride, benzene, trichloromethyl, benzyl trichloride, benzylidyne chloride, phenyltrichloromethane
PubChem CID 7367
ChEBI CHEBI:82274
IUPAC Name trichloromethylbenzene
SMILES ClC(Cl)(Cl)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -7.5°C
Color Yellow
Density 1.3800g/mL
Boiling Point 219°C to 223°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5CCl3
Refractive Index 1.5570 to 1.559
Quantity 5 mL
Beilstein 05, 300
Merck Index 15, 1112
Specific Gravity 1.38
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in alcohol,benzene,ether and,most organic solvents
Formula Weight 195.48
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material α, α, α-Trichlorotoluene, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • a,a,a-Trichlorotoluene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.