missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Trichostatin A

Product Code. 17150052
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17150052 5 mg 5mg
17140052 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17150052 Supplier STEMCELL Technologies Supplier No. 72284
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Epigenetic modifier; Inhibits histone deacetylase (HDAC)1 and HDAC6

  • Trichostatin A is a potent and reversible inhibitor of Histone Deacetylase (HDAC), therefore acting as an epigenetic modifier by preventing the removal of acetyl groups from lysine residues on histone tails
  • Trichostatin A inhibits both class I and class II HDACs, including HDAC1 (IC50 = 6 nM), HDAC4 (IC50 = 38 nM), and HDAC6 (IC50 = 8.6 nM)
  • (Furumai et al.; Yoshida et al.)

Specifications

CAS 58880-19-6
Synonym TSA
Quantity 5 mg
Molecular Formula C17H22N2O3
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.