missing translation for 'onlineSavingsMsg'
Learn More

Tricosane, 98%, Thermo Scientific Chemicals

Catalog Number 10361263
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
638-67-5
C23H48
324.63
MFCD00009350
FIGVVZUWCLSUEI-UHFFFAOYSA-N
n-tricosane, tricosan, docosane, methyl, unii-t166b8r1vc, ch3-ch2 21-ch3, tricosane, analytical standard, tricosane, tricosane standardmaterialforgc, unii-j3n6x3yk96 component
12534
CHEBI:32934
tricosane
CCCCCCCCCCCCCCCCCCCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
638-67-5
C23H48
324.63
MFCD00009350
FIGVVZUWCLSUEI-UHFFFAOYSA-N
n-tricosane, tricosan, docosane, methyl, unii-t166b8r1vc, ch3-ch2 21-ch3, tricosane, analytical standard, tricosane, tricosane standardmaterialforgc, unii-j3n6x3yk96 component
12534
CHEBI:32934
tricosane
CCCCCCCCCCCCCCCCCCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 47°C to 50°C
Color White
Boiling Point 380°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)21CH3
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 324.63
Percent Purity 98%
Physical Form Shiny Flakes
Chemical Name or Material Tricosane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'