missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ TRICOTON Light Gloves

Product Code. 12683343
Change view
Click to view available options
Size:
7
8
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12683343 7 Each
12693343 8 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 12683343 Supplier Honeywell™ Supplier No. RGT550T7
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Honeywell™ TRICOTON Light Gloves are a 100% cotton lightweight knitted glove constructed of light yarn.

  • 100% cotton lightweight knitted glove
  • The knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
  • The glove offers maximum comfort and added sturdiness
  • Of a neutral pH level and without any treatment, this cotton is tolerated perfectly well by the skin and limits the absorption of perspiration
  • The tight knit gives the glove perfect support, limits the penetration of dirt, offers a good touch while offering protection against small cuts
  • Light color enhances the glove's hygiene and cleanliness properties
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420-2003 EEC Basic standards
Construction Seamless Knit
Description 13 gauge
Material Cotton
Size 7
Glove Cuff Style Elasticated
Length (Metric) 23 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.