missing translation for 'onlineSavingsMsg'
Learn More

Tricyclohexylchlorotin, TRC

Product Code. 30454822
Change view
Click to view available options
Quantity:
2.5 g
5 g
10 g
Unit Size:
10g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454822 10 g 10g
30456829 5 g 5g
30457233 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30454822 Supplier Toronto Research Chemicals Supplier No. TRCT77450010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3091-32-5
Molecular Formula C18 H33 Cl Sn
Molecular Weight (g/mol) 403.62
Synonym Tin, chlorotricyclohexyl- (7CI), Tricyclohexyltin chloride (6CI), Chlorotricyclohexylstannane, Chlorotricyclohexyltin, NSC 202692, Tricyclohexylstannyl chloride, Tris(cyclohexyl)tin chloride
IUPAC Name chloro(tricyclohexyl)stannane
SMILES Cl[Sn](C1CCCCC1)(C2CCCCC2)C3CCCCC3

Specifications

Quantity 10 g
InChI Formula InChI=1S/3C6H11.ClH.Sn/c3*1-2-4-6-5-3-1;;/h3*1H,2-6H2;1H;/q;;;;+1/p-1
Formula Weight 404.1293
Chemical Name or Material Tricyclohexyltin chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.