missing translation for 'onlineSavingsMsg'
Learn More

Tricyclohexylphosphine, 1M solution in THF, AcroSeal™

Product Code. 10602455 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10602455 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10602455 Supplier Thermo Scientific Acros Supplier No. 427831000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2622-14-2 | C18H33P | 280.43 g/mol

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2622-14-2, 109-99-9
Color Colorless to Yellow
Density 0.9090g/mL
Flash Point −20°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C18H33P
MDL Number MFCD00003853
Quantity 100 mL
Specific Gravity 0.909
Synonym tricyclohexylphosphine, tricyclohexyl phosphine, phosphine, tricyclohexyl, tricyclohexyl-phosphane, pcy3, tricyclohexylphosphine solution, tchp, p cy 3, phosphorus tricyclohexyl, tricyclohexylphosphine, dissolved in toluene concentration
InChI Key WLPUWLXVBWGYMZ-UHFFFAOYSA-N
SMILES C1CCC(CC1)P(C2CCCCC2)C3CCCCC3
IUPAC Name tricyclohexylphosphane
Molecular Weight (g/mol) 280.43
PubChem CID 75806
Formula Weight 280.43
Physical Form Solution
Chemical Name or Material Tricyclohexylphosphine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tricyclohexylphosphine, 1M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.