missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine hydrochloride, +99%, Thermo Scientific Chemicals

Product Code. 11955271 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11955271 2.5 kg Plastic bottle 2.5kg
10582381 5 g Glass bottle 5g
10477960 100 g Glass bottle 100g
10285500 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11955271 Supplier Thermo Scientific Acros Supplier No. 170050025
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-39-8
Molecular Formula C6H16ClNO3
Molecular Weight (g/mol) 185.648
MDL Number MFCD00012596
InChI Key HHLJUSLZGFYWKW-UHFFFAOYSA-N
Synonym triethanolamine hydrochloride, tea-hydrochloride, triethanolammonium chloride, 2,2',2-nitrilotriethanol hydrochloride, ethanol, 2,2',2-nitrilotris-, hydrochloride, 2,2',2-nitrilotrisethanol hydrochloride, unii-r297uj9qdy, tris 2-hydroxyethyl ammonium chloride, tris 2-hydroxyethyl amine hydrochloride, r297uj9qdy
PubChem CID 101814
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanol;hydrochloride
SMILES C(CO)N(CCO)CCO.Cl

Specifications

Color White
Loss on Drying 1.0% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula (HOCH2CH2)3N·HCl
Quantity 2.5 kg
Formula Weight 185.65
Percent Purity ≥99.0%
Physical Form Crystalline Powder
Chemical Name or Material Triethanolamine hydrochloride, p.a.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.