missing translation for 'onlineSavingsMsg'
Learn More

Triethoxyvinylsilane, 97%, Thermo Scientific Chemicals

Product Code. 10721261 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Glass bottle
Unit Size:
25g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10721261 500 g Glass bottle 500g
10498320 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10721261 Supplier Thermo Scientific Acros Supplier No. 174615000
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-08-0
Molecular Formula C8H18O3Si
Molecular Weight (g/mol) 190.31
MDL Number MFCD00009063
InChI Key FWDBOZPQNFPOLF-UHFFFAOYSA-N
Synonym vinyltriethoxysilane, triethoxyvinylsilane, silane, ethenyltriethoxy, triethoxy vinyl silane, polyscience vtes, triethoxyvinylsilicane, vtes, silane a 151, ethenyl triethoxy silane, silane, triethoxyvinyl
PubChem CID 6516
SMILES CCO[Si](OCC)(OCC)C=C

Specifications

Color Colorless
Density 0.9000g/mL
Boiling Point 160°C to 161°C
Flash Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHSi(OC2H5)3
Refractive Index 1.3970 to 1.399
Quantity 500 g
Specific Gravity 0.9
Solubility Information Solubility in water: decomposes in water. Other solubilities: soluble in chloroform
IUPAC Name ethenyl(triethoxy)silane
Formula Weight 190.32
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Triethoxyvinylsilane, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.